Merck
  • Physical and chemical characterization of the surface layers formed on dentin following treatment with an experimental anhydrous stannous fluoride dentifrice.
Physical and chemical characterization of the surface layers formed on dentin following treatment with an experimental anhydrous stannous fluoride dentifrice.
American journal of dentistry (2013-07-10)
Jonathan S Earl, Richard M Langford
ABSTRACT
To characterize, in vitro, the mode of action of stannous fluoride containing formulations in occluding dentin tubules, by means of high resolution microscopy techniques. Focused ion beam scanning electron microscopy (FIB SEM) was used to site-specifically prepare cross sections for SEM and TEM imaging and analysis of dentin samples treated with either a stannous fluoride dispersion in glycerol or an experimental stannous fluoride dentifrice. An experimental stannous fluoride dentifrice formed a protective layer over the dentin surface and occluded dentin tubules. Additional supporting data derived from a stannous fluoride dispersion in glycerol suggest that stannous fluoride is a key component of this occluding system. Multiple SEM images obtained from sequential FIB cross-sections were reconstructed into 3-dimensional tomograms that showed a formed layer and tubule occlusion. Sections thinned by FIB techniques were observed by transmission electron microscopy (TEM) and related methods and showed that the coating, which was up to 3 microm-thick, consisted of a tin containing precipitate. Chemical analysis by energy dispersive x-ray spectroscopy (EDS) mapping that used scanning TEM (STEM) methods showed interdiffusion of tin up to 200 nm into the dentin structure.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, FCC, FG