Merck
  • [Peculiarities of pre- and postnatal kidney development in vasopressin-deficient brattleboro rats].
[Peculiarities of pre- and postnatal kidney development in vasopressin-deficient brattleboro rats].
Morfologiia (Saint Petersburg, Russia) (2013-06-29)
M S Vinogradova, A R Boiarskaia, E A Prokop'eva
ABSTRACT
The objective of this study was to examine pre- and postnatal development of the kidney in vasopressin-deficient Brattleboro rats in comparison as compared to that in Wistar rats. Histological, histochemical and morphometric methods at light microscopic level were used. The study included 50 fetuses at gestational days 16 and 18, and 46 rat pups at postnatal days 5, 10, 20, and 30. It was found that nephrogenesis sequence in both rat strains was similar, however, Brattleboro embryos and infant rats were characterized by an accelerated growth of renal corpuscles and renal tubules. The results suggest that vasopressin has no direct effect on the formation of nephron structural elements, however it may participate in the regulation of hyaluronan biosynthesis in the renal medullary interstitial tissue involved in the mechanism of urine osmotic concentration.
Sigma-Aldrich
Hyaluronic acid sodium salt from bovine vitreous humor
Sodium hyaluronate, BRP, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 10,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,200
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,200,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,500,000-1,750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,400,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 300,000-500,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 120,000-350,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 70,000-120,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 50,000-70,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 15,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,000,000-1,250,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 30,000-50,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide