Merck
Electrolytic removal of ammonia from aqueous phase by Pt/Ti anode.
Water science and technology : a journal of the International Association on Water Pollution Research (2013-06-12)
Liang Li, Yuanxing Huang, Yan Liu, Yangyang Li
ABSTRACT
This study investigated the mechanism and kinetic modeling of electrolytic degradation of ammonia with Pt/Ti anode. The results show that ammonia oxidation from direct oxidation or indirect oxidation with hydroxyl radicals was slow but can be observed under pH 9 and high initial ammonia concentration of 1,050 mg N L(-1). Indirect oxidation with HOCl was the mechanism for the chloride-mediated electrolytic removal of ammonia. In this process, pH between 3 and 9 had little effect on the ammonia removal rate, but current density (j) and chloride concentration ([Cl(-)]) showed a linear relationship with ammonia removal rate within the range of 3.8-15.4 mA cm(-2) and 30-300 mg L(-1), respectively. The ammonia removal could be described by a pseudo-zero order kinetics with a mathematic equation of k = 0.0003 × [Cl(-)] × j - 0.076. Treatment of the actual wastewater effluent from a secondary clarifier in a local wastewater treatment plant showed an ammonia removal rate of 0.8 mg N L(-1) h(-1) and energy cost of 14 kJ per mg N ammonia.
Sigma-Aldrich
Platinum, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.125-0.135 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, thermocouple grade
Sigma-Aldrich
Platinum, wire, diam. 1.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, sponge, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.20 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, gauze, 52 mesh, 99.9% trace metals basis
Sigma-Aldrich
Platinum, powder (coarse), 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.10 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, powder, 99.995% trace metals basis
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Platinum, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, black, powder, ≤20 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Platinum, gauze, 100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.127 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.25 mm, 99.99% trace metals basis