Merck
The plant cytoskeleton controls regulatory volume increase.
Biochimica et biophysica acta (2013-05-11)
Qiong Liu, Fei Qiao, Ahmed Ismail, Xiaoli Chang, Peter Nick
ABSTRACT
The ability to adjust cell volume is required for the adaptation to osmotic stress. Plant protoplasts can swell within seconds in response to hypoosmotic shock suggesting that membrane material is released from internal stores. Since the stability of plant membranes depends on submembraneous actin, we asked, whether this regulatory volume control depends on the cytoskeleton. As system we used two cell lines from grapevine which differ in their osmotic tolerance and observed that the cytoskeleton responded differently in these two cell lines. To quantify the ability for regulatory volume control, we used hydraulic conductivity (Lp) as readout and demonstrated a role of the cytoskeleton in protoplast swelling. Chelation of calcium, inhibition of calcium channels, or manipulation of membrane fluidity, did not significantly alter Lp, whereas direct manipulation of the cytoskeleton via specific chemical reagents, or indirectly, through the bacterial elicitor Harpin or activation of phospholipase D, was effective. By optochemical engineering of actin using a caged form of the phytohormone auxin we can break the symmetry of actin organisation resulting in a localised deformation of cell shape indicative of a locally increased Lp. We interpret our findings in terms of a model, where the submembraneous cytoskeleton controls the release of intracellular membrane stores during regulatory volume change.
Sigma-Aldrich
Latrunculin B from Latruncula magnifica, ≥80% (HPLC), solid
Sigma-Aldrich
Phospholipase D from Arachis hypogaea (peanut), Type II, lyophilized powder, ≥60 units/mg protein
Sigma-Aldrich
Phospholipase D from cabbage, Type IV, lyophilized powder, ≥100 units/mg solid
Sigma-Aldrich
Phospholipase D from Streptomyces sp., Type VII, lyophilized powder, ≥150 units/mg solid
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phospholipase D from Streptomyces chromofuscus, ≥50,000 units/mL, buffered aqueous glycerol solution
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard