Merck
  • Free energy, precision and learning: the role of cholinergic neuromodulation.
Free energy, precision and learning: the role of cholinergic neuromodulation.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-05-10)
Rosalyn J Moran, Pablo Campo, Mkael Symmonds, Klaas E Stephan, Raymond J Dolan, Karl J Friston
ABSTRACT
Acetylcholine (ACh) is a neuromodulatory transmitter implicated in perception and learning under uncertainty. This study combined computational simulations and pharmaco-electroencephalography in humans, to test a formulation of perceptual inference based upon the free energy principle. This formulation suggests that ACh enhances the precision of bottom-up synaptic transmission in cortical hierarchies by optimizing the gain of supragranular pyramidal cells. Simulations of a mismatch negativity paradigm predicted a rapid trial-by-trial suppression of evoked sensory prediction error (PE) responses that is attenuated by cholinergic neuromodulation. We confirmed this prediction empirically with a placebo-controlled study of cholinesterase inhibition. Furthermore, using dynamic causal modeling, we found that drug-induced differences in PE responses could be explained by gain modulation in supragranular pyramidal cells in primary sensory cortex. This suggests that ACh adaptively enhances sensory precision by boosting bottom-up signaling when stimuli are predictable, enabling the brain to respond optimally under different levels of environmental uncertainty.
Sigma-Aldrich
Acetylcholine chloride, pkg of 150 mg (per vial)
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC)
Sigma-Aldrich
Acetylcholine chloride, suitable for cell culture
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC), free-flowing, Redi-Dri
Sigma-Aldrich
Acetylcholine bromide, ≥99%
Sigma-Aldrich
Acetylcholine iodide, ≥97%
Sigma-Aldrich
Galanthamine hydrobromide from Lycoris sp., ≥94% (HPLC)