Merck
  • Stability of calcium folinate (Teva) in concentrate after re-use and in dilute infusions in 0.9% NaCl in polyethylene bags.
Stability of calcium folinate (Teva) in concentrate after re-use and in dilute infusions in 0.9% NaCl in polyethylene bags.
Acta poloniae pharmaceutica (2013-04-26)
Agnieszka Karbownik, Edyta Szałek, Hanna Urjasz, Malwina Kadziołka, Edmund Grześkowiak
ABSTRACT
The study concerned the stability of calcium folinate in concentrate in glass vials and diluted in polyethylene (PE) bags stored at 15-25 degrees C and 2-8 degrees C for up to 34 days. Original vials of calcium folinate injection (10 mg/mL, Teva) were stored at room and refrigerator temperatures and subjected to re-piercing at 1, 2, 3, 7, 14, 22, 28, 30 and 34 days following the initial piercing. Calcium folinate infusions at nominal concentrations of 0.12 mg/mL were prepared in 0.9% sodium chloride (250 mL) in PE bags. Chemical stability was measured with a stability-indicating high-performance liquid chromatography (HPLC) assay. Physical stability was assessed by visual inspection in normal light. The concentration of calcium folinate at each sampling time in the analyzed solutions remained > 90% of the initial concentration, regardless of the container. No changes in color or turbidity were observed in any of the vials or in the prepared solutions. Calcium folinate, both undiluted in glass containers and diluted with NaCl 0.9% in PE bags, remains stable (< 10% degradation) for at least 30 days at room and refrigerator temperatures when protected from light.
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Folinic acid calcium salt hydrate, BioXtra, ≥99.0% (HPLC)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, Medium density
Polyethylene (LDPE), ERM®, certified reference material