Merck
  • Inferior intrastromal corneal ring segments in paracentral keratoconus with no coincident topographic and coma axis.
Inferior intrastromal corneal ring segments in paracentral keratoconus with no coincident topographic and coma axis.
Journal of refractive surgery (Thorofare, N.J. : 1995) (2013-04-06)
José F Alfonso, Luis Fernández-Vega Cueto, Begoña Baamonde, Jesus Merayo-Lloves, David Madrid-Costa, Robert Montés-Micó
ABSTRACT
To assess the visual and refractive outcomes of implantation of intrastromal corneal ring segments (ICRS) in keratoconic eyes with no coincident topographic and comatic axes. Forty-one keratoconic eyes of 39 patients with no coincident topographic and comatic axes were implanted inferiorly with a Ferrara-type ICRS (Keraring SI6; Mediphacos Inc., Belo Horizonte, Brazil) of 150° of arc with a thickness of 150, 200, and 250 μm. Uncorrected distance visual acuity (UDVA), corrected distance visual acuity (CDVA), corneal coma-like root mean square, and residual refractive errors analyzed using vector analysis were recorded before and 6 months after the ICRS implantation. Mean UDVA was 0.76 ± 0.41 logMAR before and 0.53 ± 0.46 logMAR after surgery (P = .0006). CDVA was 0.13 ± 0.14 logMAR before and 0.07 ± 0.09 logMAR after surgery (P = .0007). Two eyes (4.9%) lost two lines or more of CDVA, 3 eyes (7.32%) lost one line, 16 eyes (39.02%) had no change in CDVA, 9 eyes (21.95%) gained one line, and 11 eyes (26.83%) gained two lines or more of CDVA. The safety index was 1.10. Spherical equivalent was significantly reduced after ICRS implantation (P < .001). Corneal coma-like root mean square changed from 0.80 ± 0.53 μm before surgery to 0.61 ± 0.59 μm after surgery (P = .02) for 4.5 mm of pupil size. One Ferrara-type ICRS of 150° of arc with a thickness of 150, 200, or 250 μm implanted inferiorly may reduce both astigmatism and corneal coma-like aberrations in keratoconic eyes with no coincident topographic and comatic axes, providing an improvement of UDVA and CDVA values.
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sigma-Aldrich
Poly(methyl methacrylate)
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,480,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 8,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 10,000
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 100,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 50,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 20,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 4,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic