Merck
  • Enamel protection from acid challenge--benefits of marketed fluoride dentifrices.
Enamel protection from acid challenge--benefits of marketed fluoride dentifrices.
The Journal of clinical dentistry (2013-04-04)
R V Faller, S L Eversole
ABSTRACT
To determine the ability of various marketed dentifrices containing stabilized stannous fluoride (SnF2), sodium fluoride (NaF), or sodium monofluorophosphate (SMFP) to protect enamel against the earliest stages of erosive dietary acid damage using an in vitro enamel protection model. Acid-challenged, extracted human teeth were treated with a 1:3 dilution of dentifrice, rinsed, and then challenged in a controlled series of tests using four dietary acids considered potentially erosive to teeth. Each acid was collected and analyzed to determine the level of mineral (phosphorous) removed from the teeth during the challenge. Post-treatment results were compared to baseline values for each acid. Results for the four acids were averaged and reported as an average percent protection value for each of the dentifrices tested, with higher values representing greater acid protection. The study included six dentifrices formulated with (A) sodium fluoride (NaF), (B) stabilized stannous fluoride (SnF2), (C,D) NaF plus 5% potassium nitrate (KNO3), (E) sodium monofluorophosphate (SMFP), or (F) SMFP plus 8% arginine bicarbonate. The stabilized SnF2 dentifrice demonstrated an average protection score of 39.3%, while products formulated with NaF resulted in protection scores between 11 and 13%. The SMFP dentifrice was rated at -3.5%, and the SMFP + arginine bicarbonate dentifrice resulted in a net average score of -5.0%. Results of this test were statistically significant (p < 0.05, ANOVA: B > A = C = D > E = F), in favor of the stabilized SnF2 dentifrice. These results suggest the stabilized SnF2 dentifrice has the potential to provide significantly better overall acid protection versus any of the other dentifrices included in the study.
Sigma-Aldrich
DL-Tartaric acid, ≥99%
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
DL-Tartaric acid, ReagentPlus®, 99%
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Potassium nitrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Mettler-Toledo Calibration substance ME 51143095, Potassium nitrate, traceable to primary standards (LGC)
Sigma-Aldrich
D-(−)-Tartaric acid, ReagentPlus®, 99%
Sigma-Aldrich
L-(+)-Tartaric acid, ≥99.5%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium fluorophosphate, 95%
Supelco
L-(+)-Tartaric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
L-(+)-Tartaric acid, ≥99.7%, FCC, FG
Sigma-Aldrich
L-(+)-Tartaric acid, puriss. p.a., reag. ISO, 99.5-101.0% (calc. to the dried substance)