Merck
  • Rapid detection of economic adulterants in fresh milk by liquid chromatography-tandem mass spectrometry.
Rapid detection of economic adulterants in fresh milk by liquid chromatography-tandem mass spectrometry.
Journal of chromatography. A (2013-04-02)
Grant Abernethy, Kerianne Higgs
ABSTRACT
A method to aid in the detection of the economically driven adulteration of fresh milk with a range of small, nitrogen containing compounds, including melamine, ammeline, ammelide, cyanuric acid, allantoin, thiourea, urea, biuret, triuret, semicarbazide, aminotriazine, 3- and 4-aminotriazole, cyanamide, dicyandiamide, guanidine, choline, hydroxyproline, nitrate, and a range of amino acids, has been developed. (15)N2-Urea is used as an internal standard. The adulteration of milk with exogenous urea has previously been difficult to detect because of the variation in the naturally occurring levels of urea in milk. However, by monitoring the contaminants biuret and triuret, which comprise up to 1% of synthetic urea, the adulteration of milk with urea-based fertilizer can be detected. We estimate that to be economically viable, adulteration of the order of 90-4000ppm of the above adulterants would need to be added to fresh milk. For most of the compounds, an arbitrary detection threshold of 2ppm is therefore more than sufficient. For biuret, a lower detection threshold, better than 0.5ppm, is desirable and the sensitivity for biuret and triuret can be improved by the post-column addition of lithium to create lithium adducts under electrospray ionisation. Sample handling involves a two-step solvent precipitation method that is deployed in a 96-well plate format, and the hydrophilic interaction liquid chromatography uses a rapid gradient (1.2min). Three separate injections, to detect the positively charged compounds, the negatively charged compounds and amino acids and finally the lithium adducts, are used. This rapid and qualitative survey method may be deployed as a second tier screening method to quickly reduce sample numbers indicated as irregular by an FTIR based screening system, and to direct analysis to appropriate quantification methods.
Sigma-Aldrich
Semicarbazide, 6 wt. % (on silica gel)
Sigma-Aldrich
Semicarbazide hydrochloride, ≥99%
Sigma-Aldrich
Guanidine sulfate salt, 99%
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Supelco
Semicarbazide hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)