Merck
Degradation performance of biodegradable Fe-Mn-C(-Pd) alloys.
Materials science & engineering. C, Materials for biological applications (2013-03-19)
Michael Schinhammer, Patrick Steiger, Frank Moszner, Jörg F Löffler, Peter J Uggowitzer
ABSTRACT
Biodegradable metals offer great potential in circumventing the long-term risks and side effects of medical implants. Austenitic Fe-Mn-C-Pd alloys feature a well-balanced combination of high strength and considerable ductility which make them attractive for use as degradable implant material. The focus of this study is the evaluation of the degradation performance of these alloys by means of immersion testing and electrochemical impedance spectroscopy in simulated body fluid. The Fe-Mn-C-Pd alloys are characterized by an increased degradation rate compared to pure Fe, as revealed by both techniques. Electrochemical measurements turned out to be a sensitive tool for investigating the degradation behavior. They not only show that the polarization resistance is a measure of corrosion tendency, but also provide information on the evolution of the degradation product layers. The mass loss data from immersion tests indicate a decreasing degradation rate for longer times due to the formation of degradation products on the sample surfaces. The results are discussed in detail in terms of the degradation mechanism of Fe-based alloys in physiological media.
Product Description
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%