Merck
  • Morphology and structural properties of high-amylose rice starch residues hydrolysed by amyloglucosidase.
Morphology and structural properties of high-amylose rice starch residues hydrolysed by amyloglucosidase.
Food chemistry (2013-03-19)
Jianmin Man, Yang Yang, Jun Huang, Changquan Zhang, Fengmin Zhang, Youping Wang, Minghong Gu, Qiaoquan Liu, Cunxu Wei
ABSTRACT
High-amylose starches are attracting considerable attention because of their potential health benefits and industrial uses. Enzyme hydrolysis of starch is involved in many biological and industrial processes. In this paper, starches were isolated from high-amylose transgenic rice (TRS) and its wild type rice, Te-qing (TQ). The morphological and structural changes of starch residues following Aspergillus niger amyloglucosidase (AAG) hydrolysis were investigated. AAG hydrolysed TQ starch from the granule surface, and TRS starch from the granule interior. During AAG hydrolysis, the content of amorphous structure increased, the contents of ordered structure and single helix decreased, and gelatinisation enthalpy decreased in TQ and TRS starch residues. The A-type polymorph of TRS C-type starch was hydrolysed faster than the B-type polymorph. The short-range ordered structure and B-type polymorph in the peripheral region of the subgranule and the surrounding band of TRS starch increased the resistance of TRS starch to AAG hydrolysis.
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Starch from corn
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from rice
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Sigma-Aldrich
Starch from corn, practical grade
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble