Merck
  • Structural basis for vitamin D receptor agonism by novel non-secosteroidal ligands.
Structural basis for vitamin D receptor agonism by novel non-secosteroidal ligands.
FEBS letters (2013-03-07)
Lisa Asano, Ichiaki Ito, Naoyuki Kuwabara, Tsuyoshi Waku, Junn Yanagisawa, Hiroyuki Miyachi, Toshiyuki Shimizu
ABSTRACT
Non-secosteroidal ligands for vitamin D receptor (VDR) have been developed for the agonist with non-calcemic profiles. Here, we provide the structural mechanism of VDR agonism by novel non-secosteroidal ligands. All ligands had the similar efficacy, while two had the higher potency. Crystallographic analyses revealed that all ligands interacted with helix H10 and the loop between helices H6 and H7 in a similar manner, but also that the two ligands with higher potency had different interaction modes. This study suggests that distinct ligand potency depend upon differences in the formation and rearrangement of hydrogen-bond networks induced by each ligand.
Product Description
Calcitriol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)