Merck
  • Mechanism of the tissue-specific action of the selective androgen receptor modulator S-101479.
Mechanism of the tissue-specific action of the selective androgen receptor modulator S-101479.
Biological & pharmaceutical bulletin (2013-03-02)
Kazuyuki Furuya, Noriko Yamamoto, Yuki Ohyabu, Teruyuki Morikyu, Hirohide Ishige, Michael Albers, Yasuhisa Endo
ABSTRACT
Selective androgen receptor modulators (SARMs) comprise a new class of molecules that induce anabolic effects with fewer side effects than those of other anabolic agents. We previously reported that the novel SARM S-101479 had a tissue-selective bone anabolic effect with diminished side effects in female animals. However, the mechanism of its tissue selectivity is not well known. In this report, we show that S-101479 increased alkaline phosphatase activity and androgen receptor (AR) transcriptional activity in osteoblastic cell lines in the same manner as the natural androgen ligand dihydrotestosterone (DHT); conversely, stimulation of AR dimerization was very low compared with that of DHT (34.4%). S-101479 increased bone mineral content in ovariectomized rats without promoting endometrial proliferation. Yeast two-hybrid interaction assays revealed that DHT promoted recruitment of numerous cofactors to AR such as TIF2, SRC1, β-catenin, NCoA3, gelsolin and PROX1 in a dose-dependent manner. SARMs induced recruitment of fewer cofactors than DHT; in particular, S-101479 failed to induce recruitment of canonical p160 coactivators such as SRC1, TIF2 and notably NCoA3 but only stimulated binding of AR to gelsolin and PROX1. The results suggest that a full capability of the AR to dimerize and to effectively and unselectively recruit all canonical cofactors is not a prerequisite for transcriptional activity in osteoblastic cells and resulting anabolic effects in bone tissues. Instead, few relevant cofactors might be sufficient to promote AR activity in these tissues.
Product Description
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Supelco
5α-Dihydrotestosterone (DHT) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®