Merck
  • Biochemical and hematologic effects of polyvinylpyrrolidone-wrapped fullerene C60 after oral administration.
Biochemical and hematologic effects of polyvinylpyrrolidone-wrapped fullerene C60 after oral administration.
Die Pharmazie (2013-03-01)
K Yamashita, Y Yoshioka, H Pan, M Taira, T Ogura, T Nagano, M Aoyama, K Nagano, Y Abe, H Kamada, S I Tsunoda, H Aoshima, H Nabeshi, T Yoshikawa, Y Tsutsumi
ABSTRACT
The fullerene C60 is used in consumer products such as cosmetics owing to its antioxidative effects and is being developed for nanomedical applications. However, knowledge regarding the safety of fullerene C60, especially after oral administration, is sparse. Here, we examined the safety of fullerene C60 in mice after 7 d of exposure to orally administered polyvinylpyrrolidone (PVP)-wrapped fullerene C60 (PVP-fullerene C60). Mice treated with PVP-fullerene C60 showed few changes in the plasma levels of various markers of kidney and liver injury and experienced no significant hematologic effects. Furthermore, the histology of the colon of PVP-fullerene C60-treated mice was indistinguishable from that of control mice. These results suggest that PVP-fullerene C60 lacks toxicity after high-dose oral administration and indicate that PVP-fullerene C60 can be considered safe for oral medication. These data provide basic information that likely will facilitate the production of safe and effective forms of fullerene C60.
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Fullerene-C60, 98%
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Fullerene-C60, sublimed, 99.9%
Sigma-Aldrich
Fullerene-C60, 99.5%
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Kollidon® 25
Sigma-Aldrich
Povidone, meets USP testing specifications
Povidone, European Pharmacopoeia (EP) Reference Standard
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material