Merck
  • Newly designed CRRT membranes for sepsis and SIRS--a pragmatic approach for bedside intensivists summarizing the more recent advances: a systematic structured review.
Newly designed CRRT membranes for sepsis and SIRS--a pragmatic approach for bedside intensivists summarizing the more recent advances: a systematic structured review.
ASAIO journal (American Society for Artificial Internal Organs : 1992) (2013-02-27)
Patrick M Honore, Rita Jacobs, Olivier Joannes-Boyau, Jouke De Regt, Elisabeth De Waele, Viola van Gorp, Willem Boer, Lies Verfaillie, Herbert D Spapen
ABSTRACT
In recent years, after all the attention has been focused on the dose for continuous renal replacement therapy (CRRT) in sepsis and systemic inflammation response syndrome (SIRS), the relatively negative results of all those studies did urge our expectations on new approaches regarding CRRT in sepsis and SIRS. So far, after the failure of the major randomized studies on dose, attention is now drawn to new membranes that could better eliminate massive amounts of unbound mediators in wider spectrum and also in greater magnitude Nevertheless, for septic acute kidney injury, the recommended dose will remain 35 ml/kg/h until the IVOIRE (hIgh VOlume in Intensive Care) study will be published. In this new armamentarium, we have distinguished the first tools that can still be called membranes ranging from AN69 Surface Treated (ST), SEPTEX, polymethylmetacrylate, to Oxiris that can still run with a CRRT device. Polymyxin B is still a kind of membrane although it has a larger surface, but it can run in a hemoperfusion system and is also much more selective. Adsorptive columns and sorbents are not anymore membranes but are seen as cartridges as the surface is extremely huge when compared with that of membranes (more than 500 m). They can still run in a hemoperfusion device. At the very end, we do have apheresis or selective plasma exchange (also very close to sorbents and columns) but we have very few data up to now regarding sepsis. Regarding spectrum, CytoSorb seems to be very promising although it is not able to capture endotoxin and IL-10. Oxiris is also promising as it can capture endotoxin and cytokines. AN69 ST is very powerful to capture numerous cytokines and especially high-mobility group box 1 protein (a very upstream cytokine). Polymethylmetacrylate has also the power to capture endotoxin and numerous other cytokines probably with a larger magnitude than Oxiris although this is not proven. Lastly, high-porosity membranes (Septex) may play a role especially when used in continuous venovenous hemodialysis mode. At the end, if we look for a more enlarged spectrum and a higher magnitude, CytoSorb might be seen as the most promising although not having the ability to fix endotoxin. Future studies will tell us which membrane or sorbent will be most useful in the adjunctive treatment for sepsis.
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 20,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 10,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 50,000
Supelco
Poly(methyl methacrylate), analytical standard, 2,480,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 8,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 100,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 4,000
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic