Merck
  • Synthesis and physicochemical properties of new tripodal amphiphiles bearing fatty acids as a hydrophobic group.
Synthesis and physicochemical properties of new tripodal amphiphiles bearing fatty acids as a hydrophobic group.
Bioorganic & medicinal chemistry letters (2013-02-16)
Prakash G Avaji, Vithal B Jadhav, Jin Xin Cui, Yong Joo Jun, Hwa Jeong Lee, Youn Soo Sohn
ABSTRACT
Saturated fatty acids (FA) were grafted using tyrosine as a spacer group to the cyclotriphosphazene ring along with equimolar hydrophilic methoxy poly(ethylene glycol) (MPEG) in cis-nongeminal way. Seven new cyclotriphosphazene amphiphiles were prepared from combinations of hydrophilic MPEGs with different molecular weights of 350, 550, 750 and 1000 and four different fatty acids of different hydrophobicity including lauric, myristic, palmitic and stearic acids. These steric amphiphiles bearing fatty acids as a hydrophobic group were found to form more stable micelles with very low critical micelle concentrations (CMC) (2.95-7.80mg/L) compared with oligopeptide analogues, and their highly hydrophobic core environment is unique and potentially useful for various biomedical applications.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 5,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, BioUltra, 500
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 550
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 750
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn ~2,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 10,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 20,000
Sigma-Aldrich
Methoxypolyethylene glycol 350, average mol wt 350