Merck
Sjogren's syndrome presenting with hypokalemic periodic paralysis.
The Journal of the Association of Physicians of India (2013-02-15)
Sumita Agrawal, Vishrant Bharti, Mayur N Jain, Prashant D Purkar, Avinash Verma, Alaka K Deshpande
ABSTRACT
We report a rare case of a 38-year-old female who presented with sudden onset flaccid quadriplegia and respiratory arrest with no significant past clinical history. She was later found to have hypokalemia due to distal renal tubular acidosis and further diagnosed as case of Sjogrens Syndrome.
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C