Merck
  • Clinical and radiological evaluation of human osseous defects (mandibular grade ii furcation involvement) treated with bioresorbable membrane: vicryl mesh.
Clinical and radiological evaluation of human osseous defects (mandibular grade ii furcation involvement) treated with bioresorbable membrane: vicryl mesh.
The journal of contemporary dental practice (2013-02-14)
K V Balusubramanya, R Ramya, S J Govindaraj
ABSTRACT
The purpose of the present study was to evaluate clinical and radiological healing effects after treatment of class II furcation defects using bioresorbable periodontal mesh barriers. The patients for the following study were selected from Outpatient, Department of Periodontics, College of Dental Sciences, Davanagere, Karnataka. 1. Patients with age group between 18 and 60 years. 2. Patients who were nonsmokers. 3. Patients who had not undergone any type of periodontal therapy 6 months prior to initial examination. 4. Patients diagnosed as advanced periodontitis having mandibular grade II furcation involvement (Glickman 1953) clinically and radiologically. 1. Patients who required antibiotic prophylaxis. 2. Patients allergic to tetracycline and/ or chlorhexidine. 3. Pregnant and lactating mother. 4. Patient showing unacceptable oral hygiene during presurgical (phase 1) therapy. In this study clinical parameters were compared and attempt was made to compare the results radiographically too, with the limitations, the present study showed that the use of resorbable periodontal mesh barriers for GTR therapy at class II furcations resulted in reduction of furcation involvement. Therefore it appears that a patient with class II furcations involvement benefits from barrier treatment, because results are superior to conventional treatment without barriers. The present study was taken up to evaluate the clinical effects of GTR therapy of class II furcations using bioresorbable periodontal mesh barriers.
Sigma-Aldrich
Resomer® RG 504 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 505, Poly(D,L-lactide-co-glycolide), ester terminated, Mw 54,000-69,000
Sigma-Aldrich
Resomer® RG 756 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 75:25, Mw 76,000-115,000
Sigma-Aldrich
Resomer® RG 502, Poly(D,L-Lactide-co-Glycolide), lactide:glycolide 50:50, ester terminated, Mw 7,000-17,000
Sigma-Aldrich
Resomer® RG 752 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 75:25, Mw 4,000-15,000
Sigma-Aldrich
Resomer® RG 653 H, Poly(D,L-lactide-co-glycolide), acid terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 503 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 502 H, Poly(D,L-lactide-co-glycolide), acid terminated, viscosity 0.16-0.24 dL/g 
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), ester terminated, Mw 50,000-75,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (75:25), mol wt 66,000-107,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (50:50), mol wt 30,000-60,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide 65:35, Mw 40,000-75,000
Sigma-Aldrich
Resomer® RG 858 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 85:15, Mw 190,000-240,000
Sigma-Aldrich
Resomer® RG 503, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 504, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 38,000-54,000