Merck
  • Magnesium sulfate during anticipated preterm birth for infant neuroprotection.
Magnesium sulfate during anticipated preterm birth for infant neuroprotection.
Nursing for women's health (2013-02-13)
Lisa Merrill
ABSTRACT
Advances in medical care of preterm infants, including the widespread use of surfactant and antenatal steroids, and improvements in ventilation management, have increased the survival rates of some of the most vulnerable infants. Yet, the risk of neurologic impairment and long-term medical complications remains a concern. Recently, the use of magnesium sulfate during anticipated preterm birth has been identified as a potential treatment to reduce adverse neurologic outcomes among preterm infants. This article discusses the use of magnesium sulfate for anticipated preterm birth to reduce neurologic impairment in preterm infants, including current clinical practice guidelines and implications for nurses.
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
Magnesium sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)