Merck
  • Influence of oxalic and malic acids in chickpea leaf exudates on the biological activity of CryIAc towards Helicoverpa armigera.
Influence of oxalic and malic acids in chickpea leaf exudates on the biological activity of CryIAc towards Helicoverpa armigera.
Journal of insect physiology (2013-02-09)
V Surekha Devi, Hari C Sharma, P Arjuna Rao
ABSTRACT
Efforts are being made to express toxin genes from the bacterium, Bacillus thuringiensis (Bt) in chickpea for minimizing the losses due to the pod borer, Helicoverpa armigera. However, there is an apprehension that acidic exudates in chickpea leaves may influence the protoxin-toxin conversion in the insect midgut, and thus, reduce the efficacy of Bt toxins. Therefore, we studied the influence of organic acids (oxalic acid and malic acid) present in the trichome exudates of chickpea on the biological activity and binding of Bt δ-endotoxin Cry1Ac to brush border membrane vesicles (BBMV) of the pod borer, H. armigera. Oxalic and malic acids in combination at concentrations present in chickpea leaves did not influence the biological activity of Bt toxin Cry1Ac towards H. armigera larvae. Amounts of Cry1Ac protein in the midgut of insects reared on diets with organic acids were similar to those reared on artificial diet without the organic acids. However, very high concentrations of the organic acids reduced the amounts of Cry1Ac in the midgut of H. armigera larvae. Organic acids in the artificial diet also increased the excretion of Cry1Ac in the fecal matter. Organic acids reduced the amount of protein in the BBMV of insects reared on diets with Cry1Ac, possibly because of reduced size of the larvae. Oxalic and malic acids at concentrations present in chickpea leaves did not affect the biological activity of Cry1Ac, but it will be desirable to have high levels of expression of Cry1Ac toxin proteins in chickpea for effective control of the pod borer, H. armigera.
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Sigma-Aldrich
L-(−)-Malic acid disodium salt, ≥95% (titration)
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Sigma-Aldrich
DL-Malic acid, ≥98% (capillary GC)
Sigma-Aldrich
Malic acid, meets USP/NF testing specifications
Sigma-Aldrich
DL-Malic acid, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
Supelco
Sodium oxalate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
D-(+)-Malic acid, unnatural form, ≥97.0% (T)
Sigma-Aldrich
DL-Malic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
DL-Malic acid, 99%
Supelco
D-Malic acid, analytical standard
Sigma-Aldrich
Sodium oxalate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium oxalate, puriss. p.a., ACS reagent, ≥99.5% (RT)
Sigma-Aldrich
Sodium oxalate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium oxalate, BioXtra