Merck
  • Streptococcus gallolyticus subspecies pasteurianus meningitis in an infant: a case report and literature review.
Streptococcus gallolyticus subspecies pasteurianus meningitis in an infant: a case report and literature review.
Journal of the Medical Association of Thailand = Chotmaihet thangphaet (2013-02-09)
Warunee Punpanich, Anurak Munsrichoom, Surang Dejsirilert
ABSTRACT
A case of 6-week-old male infant with meningitis and concurrent bacteremia caused by Streptococcus gallolyticus subspecies pasteurianus (Streptococcus bovis biotype 11.2) is presented. The isolates were susceptible to all beta-lactam antibiotics. Nevertheless, delayed defervescence and lack of satisfactory clinical improvement after treatment with multiple beta-lactam antibiotics prompted the combination therapy of vancomycin and penicillin G. The patient completed a 2-week course of antibiotics and recovered uneventfully.
Sigma-Aldrich
Penicillin G sodium salt, ~1650 U/mg
Sigma-Aldrich
Penicillin G sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Penicillin G sodium salt, 96.0-102.0%
Supelco
Penicillin G potassium salt, VETRANAL®, analytical standard
Sigma-Aldrich
Penicillin G potassium salt, 95.0 - 102.0%
Sigma-Aldrich
Penicillin G potassium salt
Sigma-Aldrich
Penicillin G potassium salt, powder, BioReagent, suitable for cell culture