Merck
  • Effect of chemical activation of 10% carbamide peroxide gel in tooth bleaching.
Effect of chemical activation of 10% carbamide peroxide gel in tooth bleaching.
The European journal of esthetic dentistry : official journal of the European Academy of Esthetic Dentistry (2013-02-08)
Graziela Ribeiro Batista, Paula Tamiao Arantes, Thomas Attin, Annette Wiegand, Carlos Rocha Torres
ABSTRACT
This study aimed to evaluate the efficacy of chemical agents to increase the bleaching effectiveness of 10% carbamide peroxide. Two hundred and ninety enamel-dentin discs were prepared from bovine incisors. The color measurement was performed by a spectrophotometer using the CIE L*a*b*system. The groups were divided according to the bleaching treatment: negative control group (NC): without bleaching; positive control group (PC): bleached with 10% carbamide peroxide gel without any chemical activator; Manganese gluconate (MG); Manganese chloride (MC); Ferrous gluconate (FG); Ferric chloride (FC); and Ferrous sulphate (FS). Three different concentrations (MG, MC, FG, FC: 0.01, 0.02 and 0.03% w/w; FS: 0.001, 0.002 and 0.003% w/w) for each agent were tested. The bleaching gel was applied on the specimens for 8 h, after which they were immersed in artificial saliva for 16 h, during 14 days. Color assessments were made after 7 and 14 days. The data were analyzed by repeated measures analysis of variance and Tukey's test (5%). Generally, the test groups were unable to increase the bleaching effect (ΔE) significantly compared to the PC group. Only for ΔL, significant higher values compared to the PC group could be seen after 7 days in groups MG (0.02%), and FS (0.002 and 0.003%). The NC group showed significantly lower values than all tested groups. It was concluded that for home bleaching procedures, the addition of chemical activators did not produce a bleaching result significantly higher than the use of 10% carbamide peroxide without activation, and that the concentration of chemical activators used did not significantly influence the effectiveness of treatment.
Sigma-Aldrich
Hydrogen peroxide–Urea adduct, powder, 15-17% active oxygen basis
Sigma-Aldrich
Iron(II) sulfate heptahydrate, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Manganese(II) chloride solution, BioReagent, for molecular biology, storage temp.:room temp
Sigma-Aldrich
D-Gluconic acid sodium salt, ≥99% (HPLC)
Sigma-Aldrich
Sodium gluconate, meets USP testing specifications
Sigma-Aldrich
Iron(II) sulfate heptahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, meets USP testing specifications
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Urea hydrogen peroxide, 97%
Sigma-Aldrich
D-Gluconic acid solution, 49-53 wt. % in H2O
Sigma-Aldrich
Manganese(II) chloride, powder and chunks, ≥99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride, beads, 98%
Supelco
D-Gluconic acid sodium salt, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ReagentPlus®, ≥99%
Supelco
Potassium gluconate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Iron(II) chloride, 98%
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Iron(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Iron(II) D-gluconate dihydrate, 98%
Sigma-Aldrich
Magnesium D-gluconate hydrate, ≥98% (HPLC)
Sigma-Aldrich
Potassium gluconate, 97.0-103.0% dry basis, meets USP testing specifications
Sigma-Aldrich
Magnesium D-gluconate hydrate, 98.0-102% anhydrous basis, meets USP testing specifications