Merck
  • Highly charged polyelectrolyte coatings to prevent adsorption during protein and peptide analysis in capillary electrophoresis.
Highly charged polyelectrolyte coatings to prevent adsorption during protein and peptide analysis in capillary electrophoresis.
Methods in molecular biology (Clifton, N.J.) (2013-02-07)
Reine Nehmé, Catherine Perrin
ABSTRACT
Capillary electrophoresis (CE) is an interesting technique for protein and peptide analysis. However, one of the major problems concerns sample adsorption on the internal capillary wall. The use of non-covalent coatings using highly charged polyelectrolytes is an efficient, simple, and fast approach to reduce peptide and protein adsorption phenomena. We have studied in a systematic manner the effect of coating conditions on the stability and efficiency of multilayer coatings using poly(diallyldimethylammonium) chloride (PDADMAC) as polycation and polystyrene sulfonate (PSS) as polyanion. When optimal conditions defined in the protocols are used, very stable coatings are obtained and adsorption phenomena are eliminated. The coatings are stable over a large range of pH buffer (2-10) and in the presence of organic solvent. Hundreds of analyses can be performed without coating regeneration. Coated capillaries can be easily stored and reused.
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 200,000-350,000 (medium molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 400,000-500,000 (high molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Poly(4-styrenesulfonic acid) solution, Mw ~75,000, 18 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw <100,000 (very low molecular weight), 35 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, 20 wt. % in H2O
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~200,000, 30 wt. % in H2O
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~1,000,000, 25 wt. % in H2O
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~70,000, 30 wt. % in H2O
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate), average Mw ~1,000,000, powder
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate), average Mw ~70,000