Merck
  • Characterization of cytosolic glutathione peroxidase and phospholipid-hydroperoxide glutathione peroxidase genes in rainbow trout (Oncorhynchus mykiss) and their modulation by in vitro selenium exposure.
Characterization of cytosolic glutathione peroxidase and phospholipid-hydroperoxide glutathione peroxidase genes in rainbow trout (Oncorhynchus mykiss) and their modulation by in vitro selenium exposure.
Aquatic toxicology (Amsterdam, Netherlands) (2013-02-07)
D Pacitti, T Wang, M M Page, S A M Martin, J Sweetman, J Feldmann, C J Secombes
ABSTRACT
Selenium (Se) is an oligonutrient with both essential biological functions and recognized harmful effects. As the selenocysteine (SeCys) amino acid, selenium is integrated in several Se-containing proteins (selenoproteins), many of which are fundamental for cell homeostasis. Nevertheless, selenium may exert toxic effects at levels marginally above those required, mainly through the generation of reactive oxygen species (ROS). The selenium chemical speciation can strongly affect the bioavailability of this metal and its impact on metabolism, dictating the levels that can be beneficial or detrimental towards an organism. Glutathione peroxidase (GPxs) is the largest and the most studied selenoprotein family. Cytosolic glutathione peroxidase (cGPx, GPx1) and phospholipid hydroperoxide glutathione peroxidase (PHGPx, GPx4) are widely distributed throughout tissues, and play a pivotal role in regulating the oxidative status in the cell. In this study we have cloned GPx1 and GPx4 genes in rainbow trout (Oncorhynchus mykiss). The constitutive mRNA expression of these GPx genes was examined in 18 trout tissues and their responsiveness to Se availability was analysed using a rainbow trout liver cell line (RTL). An inorganic (sodium selenite, Na2SeO3) and organic (selenocysteine, Cys-Se-Se-Cys) selenocompound have been used as Se sources. GPx1 activity was also tested to verify the impact of transcript changes on the enzymatic function of these molecules. To understand if the results obtained from the transcript expression analysis were due to Se bioavailability or generation of ROS, the cytoxicity of the two selenocompounds was tested by measuring the impact of Se on cell membrane integrity. Lastly, Se availability was quantified by mass spectrophotometry to determine the amount of Se in the cell culture media, the Se background due to the foetal calf serum supplement and the contribution from the two selenocompounds used in the treatments. Three isoforms of genes for both GPx1 (GPx1a, 1b1 and 1b2) and GPx4 (GPx4a1, a2 and b) have been identified. The discovery of a third gene encoding for GPx1 and GPx4 hints that salmonids may have the biggest selenoproteome amongst all vertebrates. Transcripts of GPx4 genes were more highly expressed in most tissues examined in vivo (except blood, head kidney and spleen), whereas those of the GPx1 genes were more responsive to selenium exposure in vitro, especially to the organic form. Interestingly, GPx1a was the most sensitive to selenium availability in non stressful conditions, whereas GPx1b1 and GPx1b2 were highly induced by exposure to selenium levels that had some toxic effects on the cells. Although the different concentrations tested of the two selenocompounds modulate GPx1 transcript expression to various degrees, no significant change of GPx1 enzymatic activity was detectable. Our results lead us to conclude that trout GPx1 transcripts expression level may represent a sensitive biomarker for selenium intake, helping to evaluate if selenium concentration and chemical speciation impact on cell homeostasis.
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)