Merck
  • Effects of the Ras homolog Rhes on Akt/protein kinase B and glycogen synthase kinase 3 phosphorylation in striatum.
Effects of the Ras homolog Rhes on Akt/protein kinase B and glycogen synthase kinase 3 phosphorylation in striatum.
Neuroscience (2013-02-06)
L M Harrison, S H Muller, D Spano
ABSTRACT
G protein-coupled receptors (GPCR) signal not only through heterotrimeric G proteins, but also through alternate pathways. Thus, dopamine D2 receptors in the striatum signal through Gαi/o and also by promoting formation of a multi-protein complex containing β-arrestin2, protein phosphatase 2A (PP2A), and Akt in order to dephosphorylate Akt. Lithium, on the other hand, disrupts this complex to increase Akt phosphorylation. Rhes is a striatally enriched GTP-binding protein that has been shown to inhibit dopamine receptor-mediated behavior and signaling through heterotrimeric G proteins. Therefore, our objective was to test whether Rhes similarly affects signaling through the Akt/GSK3 pathway in the striatum. Rhes(-/-) mice showed basally increased Akt and GSK3β phosphorylation relative to rhes(+/+) mice that was not further enhanced by lithium treatment. Furthermore, they responded to the D1/D2 agonist apomorphine with increased Akt and GSK3 phosphorylation. Co-immunoprecipitation experiments revealed that apomorphine treatment recruits PP 2A-C to Akt in both rhes(+/+) and rhes(-/-) mice. Lithium did not disrupt their interaction in rhes(-/-) mice as there was little basal interaction. Rhes co-immunoprecipitated with β-arrestins, suggesting that it is integral to the multi-protein complex. Thus, Rhes is necessary for Akt dephosphorylation by the striatal multi-protein complex, and in its absence, a lithium-treated phenotype results.
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride, for molecular biology, ≥99%
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithium chloride solution, 8 M, for molecular biology, ≥99%
Sigma-Aldrich
Lithium chloride, BioUltra, for molecular biology, anhydrous, ≥99.0% (AT)