Merck
  • Aerobic to anaerobic transition in Biomphalaria glabrata (Say, 1818) infected with different miracidial doses of Echinostoma paraensei (Lie and Basch, 1967) by high-performance liquid chromatography.
Aerobic to anaerobic transition in Biomphalaria glabrata (Say, 1818) infected with different miracidial doses of Echinostoma paraensei (Lie and Basch, 1967) by high-performance liquid chromatography.
Experimental parasitology (2013-02-05)
Victor Menezes Tunholi, Vinícius Menezes Tunholi-Alves, Danilo Lustrino, Rosane N Castro, Luiza D'Oliveira Sant'Ana, Juberlan Silva Garcia, Arnaldo Maldonado, Marcos Antônio José dos Santos, Maria de Lurdes de Azevedo Rodrigues, Jairo Pinheiro
ABSTRACT
The glucose content in the hemolymph and glycogen content in the digestive gland-gonad complex (DGG) and cephalopedal mass of Biomphalaria glabrata exposed to different parasite doses (5 and 50 miracidia) of Echinostoma paraensei as well as the activity of lactate dehydrogenase were evaluated. HPLC (high-performance liquid chromatography) analyses were also performed to determine the concentrations of four organic acids (oxalic, succinic, pyruvic and lactic) present in the hemolymph of infected and uninfected snails, to better understand the effect of infection on the host's energetic/oxidative metabolism. The snails were dissected 1-4 weeks after infection to collect the hemolymph and separate the tissues. There was alteration in the glycemia of the snails at both parasite doses, with a significant increase of glycemia from of the third week after infection in comparison to the control group. Changes were also observed in the lactate dehydrogenase activity, with increased activity as the infection progressed. In parallel, there was a decrease in the glycogen content in the storage tissues, with a markedly greater reduction in the digestive gland-gonad complex (larval development site) in comparison with the cephalopedal mass. Additionally, the infection by both miracidial doses resulted in an increase of oxalic and lactic acid levels, as well as in a decline of piruvic and succinic acid levels in B. glabrata, thus explaining the reduction of the oxidative decarboxylation rate in the tricarboxylic acid cycle and acceleration of the anaerobic degradation of carbohydrates in the snails, through lactic fermentation, which is essential to ensure energy supply and success of the infection.
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Sigma-Aldrich
Succinic acid, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Pyruvic acid, natural, ≥95%, FG
Sigma-Aldrich
Pyruvic acid, ≥97%, FG
Supelco
Sodium oxalate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Succinic acid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Pyruvic acid, 95%
Sigma-Aldrich
Pyruvic acid, 98%
Sigma-Aldrich
Sodium oxalate, ≥99.99% trace metals basis
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Succinic acid, puriss. p.a., ACS reagent, ≥99.5% (T)
Sigma-Aldrich
Sodium oxalate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium oxalate, puriss. p.a., ACS reagent, ≥99.5% (RT)
Sigma-Aldrich
Sodium oxalate, BioXtra
Supelco
Succinic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland