Merck
  • [Analysis of bactericidal material generated by electrical devices advertising bactericidal ability against bacteria on the agar gel plates].
[Analysis of bactericidal material generated by electrical devices advertising bactericidal ability against bacteria on the agar gel plates].
Kansenshogaku zasshi. The Journal of the Japanese Association for Infectious Diseases (2013-02-02)
Hidekazu Nishimura
ABSTRACT
Several Japanese companies sell electrical devices advertised as effective in inactivating viruses and killing bacteria by releasing special materials, e.g., Plasmacluster ions, Nanoe particle and minus ions, into the air. These companies claim that their devices killed bacteria on plates in their own experiments. We tested device effectiveness using the same experiments from the Plasmacluster ioniser SHARP Co., Japan, the Nanoe generator Panasonic Co., Japan, and the Vion KING JIM Co., Japan, to test their advertising claims. Bactericidal ability on agar plate was tested, using Staphylococcus aureus, Pseudomonas aeruginosa, Bacillus cereus, and Enterococcus faecalis as follows: the medium containing a certain amount of each bacterium was put onto an agar plate and smeared. Plates were kept in a closed chamber (inner volume 14.4 m3) or a glove box (inner volume 0.2 m), with one of the devices run for 2 hours. Plates not exposed to any device were used as controls. Each plate was retrieved and put in an incubator to count the number of bacterial colonies formed on the plate. There was no significant difference in the number of colonies on plates exposed to devices compared to control, in the number for all devices, or in all bacteria tested in experiments in the 14.4 m3 chamber. These results strongly suggest that these devices have almost no bactericidal effect, at least in space exceeding this volume. Colony formation was suppressed in the glove box in all devices and in all bacteria tested except P. aeruginosa, although the degree of suppression differed among experiments. The colony formation suppression mechanism was analyzed, and indicated that:colony formation did not change even after the removal of Plasmacluster ions, Nanoe particles, or negative ions from the air, while colony formation was decreased drastically by the removal of ozone from space, which was revealed to be generated inevitably during device operation. These results strongly suggest that the bactericidal effect seen only on the agar plate in narrow space was explained by ozone released in space as a by-product, not by special materials as advertising claimed. It is thus important to analyze the effect of special materials such as those done in this study and to suggest the involvement of ozone as the true cause, as have been done in this study, in evaluating bactericidal effect or viral inactivation as advertised by these companies.
Product Description
Millipore
Agar, powder, suitable for microbiology
Sigma-Aldrich
Agar, ash 2.0-4.5%
Millipore
Agar, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Select agar
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Agar, suitable for plant cell culture
Millipore
Agar, high purity, low ionic content, low gel strenght, suitable for microbiology
Sigma-Aldrich
Bacteriological agar, for molecular biology