Merck
  • Regiocontrolled construction of furo[3,2-c]pyran-4-one derivatives by palladium-catalyzed cyclization of propargylic carbonates with 4-hydroxy-2-pyrones.
Regiocontrolled construction of furo[3,2-c]pyran-4-one derivatives by palladium-catalyzed cyclization of propargylic carbonates with 4-hydroxy-2-pyrones.
The Journal of organic chemistry (2013-02-01)
Masahiro Yoshida, Tomomi Nakagawa, Kouki Kinoshita, Kozo Shishido
ABSTRACT
The reaction of propargylic carbonates with 4-hydroxy-2-pyrones in the presence of a palladium catalyst is described. By the choice of the reaction temperature, two types of the substituted furo[3,2-c]pyran-4-one derivatives were regioselectively synthesized, respectively.
Sigma-Aldrich
N-Methyl-N-propargylbenzylamine, 97%
Sigma-Aldrich
Pargyline hydrochloride