Merck
  • Purification of recombinant EGFP by fusion with L2 (252-273) from ribosomal protein L2 using magnetic particles.
Purification of recombinant EGFP by fusion with L2 (252-273) from ribosomal protein L2 using magnetic particles.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-01-29)
Junhua Li, Yiting Dong, Yang Zhang, Yanjun Yang
ABSTRACT
A basic polypeptide L2 (252-273) derived from Escherichia coli ribosomal protein L2 was used as a purification tag. In order to develop faster, less expensive methods for expression and purification of proteins, the L2 (252-273)-small ubiquitin like modifier (SUMO) fusion expression system was constructed. We comparatively analyzed the adsorption properties of the deleted protein of L2 (L2 (252-273)) on diatomite and superparamagnetic carboxymethyl chitosan nanoparticles. The time required to reach adsorption equilibrium of L2 (252-273) fusion protein on diatomite was shorter than that of L2 (252-273) fusion protein on magnetic particles. The maximum adsorption capacity of L2 (252-273) fusion protein on magnetic particles was about 5 times larger than that of L2 (252-273) fusion protein on diatomite. SUMO was introduced as a specific protease cleavage site between the target protein and the purification tags. The enhanced green fluorescent protein (EGFP) as a model protein was fused with the L2 (252-273)-SUMO fusion protein and purified by a simple method which involves the electrostatic adsorption of L2 (252-273) fusion proteins on superparamagnetic carboxymethyl chitosan nanoparticles and the L2 (252-273)-SUMO fusion partner was removed based on the robust cleavage by the poly lysine tagged SUMO protease. The high purity of tag-free EGFP (>93%) was obtained. Our results preliminary proved that the system was an effective fusion expression system for the production of recombinant proteins in E. coli.
Product Description
Supelco
Celatom®, acid-washed, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
Celpure® P65, meets USP/NF testing specifications
Sigma-Aldrich
Celite® 503, filter aid, treated with sodium carbonate, calcined
Supelco
Celite® 545 AW, reagent grade
Sigma-Aldrich
Hyflo® Super Cel®, filter aid, flux calcined, treated with sodium carbonate
Sigma-Aldrich
Diatomaceous earth non-washed, powder
Sigma-Aldrich
Filter agent, Celite® 545
Sigma-Aldrich
Diatomaceous earth
Sigma-Aldrich
Celpure® P1000, meets USP/NF testing specifications
Sigma-Aldrich
Celite® 545 AW, filter aid, acid washed, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Celite® S, filter aid, dried, untreated
Sigma-Aldrich
Celite® 545, filter aid, treated with sodium carbonate, flux calcined