Merck
  • Antifungal activity of volatile organic compounds from Streptomyces alboflavus TD-1.
Antifungal activity of volatile organic compounds from Streptomyces alboflavus TD-1.
FEMS microbiology letters (2013-01-29)
Changlu Wang, Zhifang Wang, Xi Qiao, Zhenjing Li, Fengjuan Li, Mianhua Chen, Yurong Wang, Yufang Huang, Haiyan Cui
ABSTRACT
Streptomyces sp. TD-1 was identified as Streptomyces alboflavus based on its morphological characteristics, physiological properties, and 16S rDNA gene sequence analysis. The antifungal activity of the volatile-producing S. alboflavus TD-1 was investigated. Results showed that volatiles generated by S. alboflavus TD-1 inhibited storage fungi Fusarium moniliforme Sheldon, Aspergillus flavus, Aspergillus ochraceus, Aspergillus niger, and Penicillum citrinum in vitro. GC/MS analysis revealed that 27 kinds of volatile organic compounds were identified from the volatiles of S. alboflavus TD-1 mycelia, among which the most abundant compound was 2-methylisoborneol. Dimethyl disulfide was proved to have antifungal activity against F. moniliforme by fumigation in vitro.
Sigma-Aldrich
2-Methylisoborneol, ≥98.0% (GC)
Sigma-Aldrich
Dimethyl disulfide, ≥99%
Sigma-Aldrich
Dimethyl disulfide, ≥98%, FG
Sigma-Aldrich
2-Methylisoborneol solution, ~10 mg/mL in methanol, ≥98% (GC)
Sigma-Aldrich
Dimethyl disulfide, ≥99.0%