Merck
  • A multifunctional biphasic suspension of mesoporous silica encapsulated with YVO4:Eu3+ and Fe3O4 nanoparticles: synergistic effect towards cancer therapy and imaging.
A multifunctional biphasic suspension of mesoporous silica encapsulated with YVO4:Eu3+ and Fe3O4 nanoparticles: synergistic effect towards cancer therapy and imaging.
Nanotechnology (2013-01-18)
N Shanta Singh, Hrishikesh Kulkarni, Lina Pradhan, D Bahadur
ABSTRACT
Polyol mediated synthesized luminescent YVO(4):Eu(3+) nanoparticles (NPs) have been encapsulated in mesoporous silica nanoparticles (MSNs) using the sol-gel process. X-ray diffraction and Fourier transform infrared spectroscopy along with transmission electron microscopy confirm the encapsulation of the YVO(4):Eu(3+) NPs in the SiO(2) matrix. N(2) adsorption/desorption analysis confirms the mesoporous nature of the MSNs and YVO(4):Eu(3+)-MSNs. No significant quenching of the YVO(4):Eu(3+) luminescence is observed for YVO(4):Eu(3+)-MSNs. This nanocomposite has been tested as a potential drug carrier. Efficient loading of doxorubicin hydrochloride (DOX), a typical anticancer drug, is observed which reaches up to 93% in 8 mg ml(-1) of YVO(4):Eu(3+)-MSNs. pH sensitive release of DOX is observed, with 54% release for pH 4.3 and 31% in a physiological environment (pH 7.4). Both MSNs and YVO(4):Eu(3+)-MSNs nanocomposites do not show accountable toxicity to two cell lines, i.e. HeLa and MCF-7. However, as desired, toxicity is observed when cells are incubated with DOX loaded YVO(4):Eu(3+)-MSNs. Laser scanning confocal microscopy images confirm the uptake of the nanocomposite in both cell lines. The morphology of the cells (MCF-7) changes after incubation with DOX loaded YVO(4):Eu(3+)-MSNs, indicating an interaction of DOX with the cells. More cytotoxicity to both cell lines with ∼90% killing is observed due to the synergistic effect of magnetic fluid hyperthermia and chemotherapy using a biphasic suspension of superparamagnetic iron oxide magnetic nanoparticles and DOX loaded YVO(4):Eu(3+)-MSNs. In addition, an AC magnetic field triggers an enhanced drug release.
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron(II,III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Iron(II,III) oxide, powder, <5 μm, 95%
Sigma-Aldrich
Iron(II,III) oxide, nanopowder, 50-100 nm particle size (SEM), 97% trace metals basis
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Yttrium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Yttrium sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis