Merck
  • A simple system for expression of proteins containing 3-azidotyrosine at a pre-determined site in Escherichia coli.
A simple system for expression of proteins containing 3-azidotyrosine at a pre-determined site in Escherichia coli.
Journal of biochemistry (2013-01-15)
Akiyoshi Ikeda-Boku, Satoshi Ohno, Yuuka Hibino, Takashi Yokogawa, Nobuhiro Hayashi, Kazuya Nishikawa
ABSTRACT
We developed an efficient method for introduction of 3-azidotyrosine (N(3)-Y) into proteins in Escherichia coli cells. We constructed a plasmid that is adaptable for the constitutive expression of both Methanosarcina acetivorans tyrosyl-tRNA synthetase (TyrRS) and tRNA(()(CUA)), and made an orthogonal tRNA((CUA)) that is recognized as a substrate only by the archaeal TyrRS. Random mutations were introduced into M. acetivorans TyrRS around the tyrosine binding pocket, and a TyrRS mutant recognizing N(3)-Y was selected. We then expressed rat calmodulin (CaM) containing N(3)-Y, using the CaM gene with an amber codon at position 80. Mass analyses confirmed production of CaM containing N(3)-Y, but a significant amount of CaM containing 3-aminotyrosine was also detected. To more efficiently express CaM containing N(3)-Y, we added an arabinose-inducible gene for the mutant TyrRS to the plasmid carrying the mutant TyrRS/tRNA(()(CUA)) gene. Although the yields of full-length CaM increased ~3-fold, the ratio of N(3)-Y introduction was not significantly improved. Following screening for a suitable host cell, we found that CaM expressed in E. coli SHuffle (K-12) had 97% N(3)-Y at the pre-determined site. Finally, we obtained up to 2 mg of CaM containing N(3)-Y per 100 ml of culture media, sufficient for use in various proteomics experiments, including photo-crosslinking.
Sigma-Aldrich
DL-Arabinose, ≥98% (GC)