Merck
  • Metabolomic profilings of urine and serum from high fat-fed rats via 1H NMR spectroscopy and pattern recognition.
Metabolomic profilings of urine and serum from high fat-fed rats via 1H NMR spectroscopy and pattern recognition.
Applied biochemistry and biotechnology (2013-01-12)
Jingjing Xu, Changqin Liu, Shuhui Cai, Jiyang Dong, Xuejun Li, Jianghua Feng, Zhong Chen
ABSTRACT
(1)H NMR spectroscopy in combination with multivariate statistical analysis was applied to explore the metabolic variability in urine and serum of high fat-fed rats relative to normal chow-fed ones. Metabolites contributing to intergroup discrimination identified by partial least squares discriminant analysis include 3-hydroxybutyrate, glutamate, glutamine, citrate, choline, hippurate, alanine, lactate, creatinine, taurine, acetate, etc. The aging effect along with long-term feeding was delineated with metabolic trajectory in principal component analysis score plot and age-related differences on metabolic profiling under different dietary intervention were recognised. The identified metabolites responsible for obesity were all imported into a web tool for network-based interpretation of compound lists to interpret their functional context, molecular mechanisms and disturbed signalling pathway globally and systematically. The results are useful for interpreting the pathology of obesity and further probing into the relationship between dietary-induced obesity and type 2 diabetes mellitus.
Sigma-Aldrich
Sodium hippurate hydrate, ≥99%
Sigma-Aldrich
Hippuric acid, 98%
Sigma-Aldrich
3-Hydroxybutyric acid, 95%