Merck
  • ASR1 mediates glucose-hormone cross talk by affecting sugar trafficking in tobacco plants.
ASR1 mediates glucose-hormone cross talk by affecting sugar trafficking in tobacco plants.
Plant physiology (2013-01-11)
Pia Guadalupe Dominguez, Nicolas Frankel, Jeannine Mazuch, Ilse Balbo, Norberto Iusem, Alisdair R Fernie, Fernando Carrari
ABSTRACT
Asr (for ABA, stress, ripening) genes are exclusively found in the genomes of higher plants, and the encoded proteins have been found localized both to the nucleus and cytoplasm. However, before the mechanisms underlying the activity of ASR proteins can be determined, the role of these proteins in planta should be deciphered. Results from this study suggest that ASR is positioned within the signaling cascade of interactions among glucose, abscisic acid, and gibberellins. Tobacco (Nicotiana tabacum) transgenic lines with reduced levels of ASR protein showed impaired glucose metabolism and altered abscisic acid and gibberellin levels. These changes were associated with dwarfism, reduced carbon dioxide assimilation, and accelerated leaf senescence as a consequence of a fine regulation exerted by ASR to the glucose metabolism. This regulation resulted in an impact on glucose signaling mediated by Hexokinase1 and Snf1-related kinase, which would subsequently have been responsible for photosynthesis, leaf senescence, and hormone level alterations. It thus can be postulated that ASR is not only involved in the control of hexose uptake in heterotrophic organs, as we have previously reported, but also in the control of carbon fixation by the leaves mediated by a similar mechanism.
Sigma-Aldrich
Gibberellic acid, 90% gibberellin A3 basis (HPLC)
Supelco
Gibberellic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Gibberellic acid potassium salt, BioReagent, suitable for plant cell culture, ~95%, ≥50% total GA3 basis
Sigma-Aldrich
Gibberellic acid, BioReagent, suitable for plant cell culture, ≥90% gibberellin A3 basis (of total gibberellins.)