Merck
  • Pectin and charge modified pectin hydrogel beads as a colon-targeted drug delivery carrier.
Pectin and charge modified pectin hydrogel beads as a colon-targeted drug delivery carrier.
Colloids and surfaces. B, Biointerfaces (2013-01-10)
Jiyoung Jung, Robert D Arnold, Louise Wicker
ABSTRACT
The physical and chemical properties of commercial low methoxyl citrus pectins, CP 28 and CP 55, and a pectinmethylesterase (PME) charge modified citrus pectin (MP 38) were compared, and the differences in ability to encapsulate indomethacin in hydrogel beads was determined at 0.5 or 1.0% (w/v) indomethacin ratio, and 100, 200 or 300 mM CaCl(2) solution. In order to investigate the drug release characteristics, indomethacin loaded dried hydrogel beads were immersed in simulated gastric fluids (pH 1.2) for 2h, followed by immersing in simulated intestinal fluids (pH 7.4) for 3h. Pectin type was highly significant (p<0.0001) for encapsulation efficiency and in vitro release assay. Encapsulation efficiency was also highly affected (p<0.0001) by indomethacin ratio and CaCl(2) concentration. The accumulative release rate of indomethacin from pectin hydrogel bead was less than 15% in simulated gastro-intestinal fluids. MP 38 beads showed significantly higher entrapment efficiency and lower release rate than beads formed from CP 28 or CP 55. MP 38 hydrogel formulated with 300 mM CaCl(2) and 0.5% indomethacin ratio showed the highest entrapment efficiency. These studies suggest that charge modification of pectin improves encapsulation efficiency of drugs for colon targeted drug delivery system through oral administration.
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Pectin from citrus peel, Galacturonic acid ≥74.0 % (dried basis)
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, for molecular biology, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Supelco
Pectin, for use with Total Dietary Fiber Control Kit, TDF-C10
Supelco
Calcium ion solution for ISE, 0.1 M Ca, for ion-selective electrodes, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%