Merck
  • The multigene family of lysophosphatidate acyltransferase (LPAT)-related enzymes in Ricinus communis: cloning and molecular characterization of two LPAT genes that are expressed in castor seeds.
The multigene family of lysophosphatidate acyltransferase (LPAT)-related enzymes in Ricinus communis: cloning and molecular characterization of two LPAT genes that are expressed in castor seeds.
Plant science : an international journal of experimental plant biology (2012-12-26)
José María Arroyo-Caro, Tarik Chileh, Michael Kazachkov, Jitao Zou, Diego López Alonso, Federico García-Maroto
ABSTRACT
The multigene family encoding proteins related to lysophosphatidyl-acyltransferases (LPATs) has been analyzed in the castor plant Ricinus communis. Among them, two genes designated RcLPAT2 and RcLPATB, encoding proteins with LPAT activity and expressed in the developing seed, have been cloned and characterized in some detail. RcLPAT2 groups with well characterized members of the so-called A-class LPATs and it shows a generalized expression pattern in the plant and along seed development. Enzymatic assays of RcLPAT2 indicate a preference for ricinoleoyl-CoA over other fatty acid thioesters when ricinoleoyl-LPA is used as the acyl acceptor, while oleoyl-CoA is the preferred substrate when oleoyl-LPA is employed. RcLPATB groups with B-class LPAT enzymes described as seed specific and selective for unusual fatty acids. However, RcLPATB exhibit a broad specificity on the acyl-CoAs, with saturated fatty acids (12:0-16:0) being the preferred substrates. RcLPATB is upregulated coinciding with seed triacylglycerol accumulation, but its expression is not restricted to the seed. These results are discussed in the light of a possible role for LPAT isoenzymes in the channelling of ricinoleic acid into castor bean triacylglycerol.
Sigma-Aldrich
Castor oil
Sigma-Aldrich
Castor oil, tested according to Ph. Eur.
Sigma-Aldrich
Ricinoleic acid, ≥95%
Sigma-Aldrich
Castor oil, meets testing specifications per USP