Merck
  • Chemoselective acylation of (hydroxyalkyl)phenols catalyzed by Candida antarctica lipase B.
Chemoselective acylation of (hydroxyalkyl)phenols catalyzed by Candida antarctica lipase B.
Biotechnology letters (2012-12-20)
Toshifumi Miyazawa, Masato Yamamoto, Hiroshi Danjo
ABSTRACT
Acylation of (hydroxyalkyl)phenols with vinyl esters by lipase B from Candida antarctica proceeded smoothly in a highly chemoselective manner, affording their alkyl esters exclusively or at least predominantly. The enzyme therefore discriminates between an alcoholic hydroxyl from a phenolic hydroxyl in addition to having versatile catalytic abilities for organic synthesis.
Product Description
Supelco
Vinyl acetate, analytical standard
Sigma-Aldrich
Vinyl acetate, contains 3-20 ppm hydroquinone as inhibitor, ≥99%
Vinyl acetate, European Pharmacopoeia (EP) Reference Standard
Supelco
Vinyl acetate solution, certified reference material, 5000 μg/mL in acetonitrile