Merck
  • Terpenoids of plant origin inhibit morphogenesis, adhesion, and biofilm formation by Candida albicans.
Terpenoids of plant origin inhibit morphogenesis, adhesion, and biofilm formation by Candida albicans.
Biofouling (2012-12-12)
Jayant S Raut, Ravikumar B Shinde, Nitin M Chauhan, S Mohan Karuppayil
ABSTRACT
Biofilm-related infections caused by Candida albicans and associated drug resistant micro-organisms are serious problems for immunocompromised populations. Molecules which can prevent or remove biofilms are needed. Twenty-eight terpenoids of plant origin were analysed for their activity against growth, virulence attributes, and biofilms of C. albicans. Eighteen molecules exhibited minimum inhibitory concentrations of <2 mg ml(-1) for planktonic growth. Selected molecules inhibited yeast to hyphal dimorphism at low concentrations (0.031-0.5 mg ml(-1)), while adhesion to a solid surface was prevented at 0.5-2 mg ml(-1). Treatment with 14 terpenoids resulted in significant (p < 0.05) inhibition of biofilm formation, and of these, linalool, nerol, isopulegol, menthol, carvone, α-thujone, and farnesol exhibited biofilm-specific activity. Eight terpenoids were identified as inhibitors of mature biofilms. This study demonstrated the antibiofilm potential of terpenoids, which need to be further explored as therapeutic strategy against biofilm associated infections of C. albicans.
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Crystal Violet, S. No.: 785
Sigma-Aldrich
Crystal Violet
Sigma-Aldrich
Crystal Violet, certified by the BSC
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis