Merck
  • Biodegradation of di-n-butyl phthalate by a stable bacterial consortium, HD-1, enriched from activated sludge.
Biodegradation of di-n-butyl phthalate by a stable bacterial consortium, HD-1, enriched from activated sludge.
Bioresource technology (2012-12-04)
Zhixing He, Hailong Xiao, Lu Tang, Hang Min, Zhenmei Lu
ABSTRACT
HD-1, a stable microbial consortium capable of mineralizing di-n-butyl phthalate (DBP), was developed from activated sludge. The dominant microorganisms in the consortium, Gordonia sp., Burkholderia sp. and Achromobacter sp., were identified by denaturing gradient gel electrophoresis (DGGE). The consortium could mineralize approximately 90% of 1200 mg/L DBP after 48 h of cultivation. The optimal DBP degradation conditions were 25-30 °C and pH 8.0-9.0. The addition of yeast (0.5 g/L), sodium acetate (0.5 g/L, 1.0 g/L), Brij 35 (0.2%, 0.5%, 1.0%), or Triton X-100 (0.2%) enhanced DBP degradation. The DBP degradation rate was influenced by the presence of dimethyl phthalate (DMP) and diethyl phthalate (DEP). Only one main intermediate, phthalic acid, could be monitored by gas chromatography-mass spectrometry (GC-MS) during the degradation process. The HD-1 consortium also utilized phenol, o-dihydroxybenzene as the sole carbon and energy source. The results indicate the consortium may represent a promising application for DBP bioremediation.
Sigma-Aldrich
Dibutyl phthalate, 99%
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Supelco
Di-n-butyl phthalate solution, certified reference material, 5000 μg/mL in methanol
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland