Merck
  • Uptake of gold (III) from waste gold solution onto biomass-based adsorbents organophosphonic acid functionalized spent buckwheat hulls.
Uptake of gold (III) from waste gold solution onto biomass-based adsorbents organophosphonic acid functionalized spent buckwheat hulls.
Bioresource technology (2012-12-01)
Ping Yin, Mingyu Xu, Rongjun Qu, Hou Chen, Xiguang Liu, Jiang Zhang, Qiang Xu
ABSTRACT
Novel biomass-based adsorbents organophosphonic acid functionalized spent buckwheat hulls (OPA-BH) with 60 mesh were successfully employed to adsorb Au(III) ions from simulated wastewater. The adsorption kinetics and isotherms both in unary ion system and in ternary ions system were investigated, and the applicability of the Langmuir, Freundlich and extended Langmuir isotherm models has been tested for the equilibrium. The process optimization was also conducted by using response surface methodology (RSM), and the maximum adsorption capacities reached 2.84 ± 0.01 mmol/g under the optimum process conditions. Furthermore, the regeneration capacities of OPA-BH were investigated by using the eluent solutions of 0.0-5.0% thiourea in 0.1 mmol/L HCl, and the results showed that the adsorption capabilities for OPA-BH were ranged between 0.77 mmol/g and 0.85 mmol/g after three cycles of adsorption-desorption processes. The research results showed that OPA-BH was favorable and useful for gold adsorption, and the high adsorption capacity and good reproducibility make it a good promising material for the precious metal uptake.
Sigma-Aldrich
Etidronate disodium hydrate, ≥97% (NMR), solid
Sigma-Aldrich
Etidronic acid, 60% aqueous solution