Merck
  • Manganese-induced effects on testicular trace element levels and crucial hormonal parameters of Hyline cocks.
Manganese-induced effects on testicular trace element levels and crucial hormonal parameters of Hyline cocks.
Biological trace element research (2012-11-30)
Xiao-fei Liu, Li-ming Zhang, Ziwei Zhang, Ning Liu, Shi-wen Xu, Hong-jin Lin
ABSTRACT
Manganese (Mn) is an essential element required for normal development and reproduction. However, little is known about the reproductive toxicity of Mn in birds. To investigate the Mn-induced toxicity on testicular trace element levels and crucial hormonal parameters on male reproduction in birds, 50-day-old male Hyline cocks were fed either a commercial diet or a Mn-supplemented diet. The changes in contents of copper (Cu), iron (Fe), zinc (Zn), and calcium (Ca) in testis were detected. Hormonal parameters were evaluated including the levels of testosterone (T), luteinizing hormone (LH), follicle-stimulating hormone (FSH), thyroid-stimulating hormone (TSH), triiodothyronine (T3), and thyroxine (T4) in the serum. The mRNA levels of luteinizing hormone receptor (LHR) and follicle-stimulating hormone receptor (FSHR) were determined in this study. The results showed that Mn was accumulated in testis, and the content of Cu, Fe, Zn, and Ca decreased. Exposure to Mn significantly lowered the content of T, LH, FSH, and the mRNA expression levels of LHR and FSHR. Levels of T3 and T4 appeared with a decreased tendency, and TSH presented no obvious regularity. It indicated that Mn exposure resulted in the disbalance of testicular trace elements and influenced hormone levels in the molecular level, which may be possible underlying reproductive toxicity mechanism induced by Mn.
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Manganese(II) chloride, powder and chunks, ≥99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride, beads, 98%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, meets USP testing specifications
Sigma-Aldrich
Manganese(II) chloride solution, BioReagent, for molecular biology, storage temp.:room temp
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride 0.1 M solution