Merck
  • Fate of deoxynivalenol, T-2 and HT-2 toxins and their glucoside conjugates from flour to bread: an investigation by high-performance liquid chromatography high-resolution mass spectrometry.
Fate of deoxynivalenol, T-2 and HT-2 toxins and their glucoside conjugates from flour to bread: an investigation by high-performance liquid chromatography high-resolution mass spectrometry.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2012-11-20)
Elisabetta De Angelis, Linda Monaci, Michelangelo Pascale, Angelo Visconti
ABSTRACT
Deoxynivalenol, T-2 and HT-2 toxins are mycotoxins frequently occurring in cereals and cereal-based products along with their conjugated forms. In this paper, we provide insights into the fate of deoxynivalenol, T-2 and HT-2 toxins and their glucoside derivatives during bread making, using naturally contaminated wheat flour. High-resolution mass spectrometry was used to assess the extent of degradation of the three mycotoxins during bread baking and to identify some glucoside conjugates, namely deoxynivalenol, T-2 and HT-2 mono-glucosides, detected both in the flour and in the respective breads. Our findings show deoxynivalenol's levels markedly increased upon baking, whereas those of HT-2 and T-2 toxins were decreased in the final bread with special regard to the T-2 toxin.
Sigma-Aldrich
HT-2 Toxin, ≥98% (HPLC)
Sigma-Aldrich
T-2 Toxin, from Fusarium sp., powder, ≥98% (HPLC)
Supelco
T2-Toxin solution, 100 μg/mL in acetonitrile, analytical standard