Merck
  • Control of the electrostatic effect on DAM-adsorbent for the water-soluble compounds by HPLC.
Control of the electrostatic effect on DAM-adsorbent for the water-soluble compounds by HPLC.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2012-11-15)
Waka Kamichatani, Yoshinori Inoue, Atsushi Yamamoto
ABSTRACT
The effect of the mobile phase pH on the control of the electrostatic interaction was evaluated on a column packed with water-holding adsorbent on which diallylamine-maleic acid copolymers were immobilizing. The adsorbent showed extraordinary retention behaviors of water-soluble solutes under acidic conditions, however, their behavior became stable along with increasing pH. Hydrating water contents tended to level off at pH above 8. Thus, the electrostatic interaction with the stationary phase can be controlled by adjusting the mobile phase pH above 8. In this region, the retention of water-soluble solutes appears to be mainly governed by the hydrophilic partition interaction.
Product Description
Supelco
Maleic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Maleic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium maleate dibasic, purum p.a., anhydrous, ≥98.0% (NT)
Sigma-Aldrich
Maleic acid sodium salt, ~99%