Merck
Investigating silver coordination to mixed chalcogen ligands.
Molecules (Basel, Switzerland) (2012-11-10)
Fergus R Knight, Rebecca A M Randall, Lucy Wakefield, Alexandra M Z Slawin, J Derek Woollins
ABSTRACT
Six silver(I) coordination complexes have been prepared and structurally characterised. Mixed chalcogen-donor acenaphthene ligands L1-L3 [Acenap(EPh)(E'Ph)] (Acenap = acenaphthene-5,6-diyl; E/E' = S, Se, Te) were independently treated with silver(I) salts (AgBF₄/AgOTf). In order to keep the number of variables to a minimum, all reactions were carried out using a 1:1 ratio of Ag/L and run in dichloromethane. The nature of the donor atoms, the coordinating ability of the respective counter-anion and the type of solvent used in recrystallisation, all affect the structural architecture of the final silver(I) complex, generating monomeric, silver(I) complexes {[AgBF₄(L)₂] (1 L = L1; 2 L = L2; 3 L = L3), [AgOTf(L)₃] (4 L = L1; 5 L = L3), [AgBF₄(L)₃] (2a L = L1; 3a L = L3)} and a 1D polymeric chain {[AgOTf(L3)](n) 6}. The organic acenaphthene ligands L1-L3 adopt a number of ligation modes (bis-monodentate μ₂-η²-bridging, quasi-chelating combining monodentate and η⁶-E(phenyl)-Ag(I) and classical monodentate coordination) with the central silver atom at the centre of a tetrahedral or trigonal planar coordination geometry in each case. The importance of weak interactions in the formation of metal-organic structures is also highlighted by the number of short non-covalent contacts present within each complex.
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis
Sigma-Aldrich
Zinc trifluoromethanesulfonate, 98%
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 99.995% trace metals basis
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 96%
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99%
Sigma-Aldrich
Silver tetrafluoroborate, ≥99.99% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99.95% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, purum, ≥98.0% (Ag)
Sigma-Aldrich
Silver tetrafluoroborate, 98%
Sigma-Aldrich
Aluminum trifluoromethanesulfonate, 99.9% trace metals basis