Merck
Amylose folding under the influence of lipids.
Carbohydrate research (2012-11-07)
Cesar A López, Alex H de Vries, Siewert J Marrink
ABSTRACT
The molecular dynamics simulation technique was used to study the folding and complexation process of a short amylose fragment in the presence of lipids. In aqueous solution, the amylose chain remains as an extended left-handed helix. After the addition of lipids in the system, however, we observe spontaneous folding of the amylose chain into a helical structure, with helical pitch and hydrogen bond network compatible with the V-amylose structure observed in X-ray experiments. Our results suggest that under the influence of external non polar ligands, the conformation of amylose undergoes a transition from an extended to a V-amylose structure in line with experimental evidence.
Product Description
Glycerol mono-oleate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nonane, anhydrous, ≥99%
Sigma-Aldrich
Nonane, ReagentPlus®, 99%