Merck
  • Cost-effectiveness of perioperative selective decontamination of the digestive tract versus placebo in elective gastrointestinal surgery.
Cost-effectiveness of perioperative selective decontamination of the digestive tract versus placebo in elective gastrointestinal surgery.
Digestive surgery (2012-11-07)
L M Dijksman, D Roos, M F Gerhards, J G P Tijssen, D J Gouma, M G W Dijkgraaf
ABSTRACT
We compared the routine use of perioperative selective decontamination of the digestive tract (SDD) for elective gastrointestinal surgery with placebo in a randomized controlled trial. Alongside this trial, a cost-effectiveness analysis from a provider perspective was performed. A total of 289 patients undergoing elective surgery of the digestive tract were randomized to either SDD (143 patients) or placebo (146 patients). Routine use of SDD led to less patients with an infectious complication compared with placebo (p = 0.028). Mean total costs per patient were slightly less (EUR 2,604; 95% CI -6,292 to 1,084) in patients randomized to SDD (EUR 12,031) compared to patients randomized to placebo (EUR 14,635). Costs of hospitalization were the main determinant of the cost difference between the groups. The incremental cost-effectiveness ratio per prevented occurrence of ≥1 infectious complications per patient was -EUR 23,164, indicating the superiority of SDD over placebo. This study shows that in patients undergoing elective gastrointestinal surgery, the routine use of SDD is less expensive and economically more efficient than placebo in reducing the number of patients with infectious complications.
Sigma-Aldrich
Polymyxin B solution, 20 mg/mL in H2O
Sigma-Aldrich
Polymyxin B sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Polymyxin B sulfate salt
Sigma-Aldrich
Tobramycin, Aminoglycoside antibiotic
Supelco
Polymyxin B solution, 1 mg/mL in H2O, analytical standard
Supelco
Tobramycin, Pharmaceutical Secondary Standard; Certified Reference Material
Polymyxin B sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Polymyxin B sulfate, meets USP testing specifications