Merck
  • [Liraglutide: new results in the treatment of type 2 diabetes mellitus].
[Liraglutide: new results in the treatment of type 2 diabetes mellitus].
Drugs of today (Barcelona, Spain : 1998) (2012-10-20)
J L Mateos, B L Wajchenberg
ABSTRACT
New drugs for type 2 diabetes that act on incretin metabolism have been shown to improve glycemic control, reduce body weight and have a low risk for hypoglycemia. Among these, liraglutide is the first glucagon-like peptide 1 (GLP-1) analogue approved for subcutaneous, once-daily administration. According to results from clinical trials, liraglutide is an attractive alternative for the early treatment of type 2 diabetes. The results of the LEAD (Liraglutide Effect and Action in Diabetes) study program demonstrated the efficacy and safety of liraglutide in terms of reduction of glycated hemoglobin (HbA1c) levels, significant loss of body weight that was maintained over the long term, better control of the lipid profile and systolic arterial pressure, reduction of the risk for hypoglycemia and reduction of cardiovascular risk. Moreover, the drug was demonstrated to be safe and can be co-administered with oral antidiabetic agents. The product's tolerability has been demonstrated, with nausea as the most common adverse event, which waned from the fourth week of treatment.
Product Description
Glimepiride for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glimepiride, ≥98% (HPLC), solid