Merck
  • Rapid identification of carbapenemase types in Enterobacteriaceae and Pseudomonas spp. by using a biochemical test.
Rapid identification of carbapenemase types in Enterobacteriaceae and Pseudomonas spp. by using a biochemical test.
Antimicrobial agents and chemotherapy (2012-10-17)
Laurent Dortet, Laurent Poirel, Patrice Nordmann
ABSTRACT
A biochemical test (Carba NP test II) was developed to identify carbapenemase production in Enterobacteriaceae and Pseudomonas spp. and to discriminate between the different types of carbapenemases (classes A, B, and D). It is based on the detection of the acidification resulting from imipenem hydrolysis, coupled with tazobactam and EDTA as inhibitors. This is an easy and reliable technique (100% sensitivity and specificity) for detection of not only carbapenemase activity but also carbapenemase types in Enterobacteriaceae and Pseudomonas aeruginosa.
Sigma-Aldrich
Zinc sulfate solution, 0.3 N
Sigma-Aldrich
Zinc sulfate heptahydrate, meets USP testing specifications
Sigma-Aldrich
Zinc sulfate heptahydrate, suitable for plant cell culture
Sigma-Aldrich
Zinc sulfate heptahydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Zinc sulfate heptahydrate, BioUltra, for molecular biology, 2.0 M in H2O
Sigma-Aldrich
Zinc sulfate heptahydrate, ACS reagent, 99%
Sigma-Aldrich
Zinc sulfate heptahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Zinc sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Zinc sulfate heptahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%