Merck
  • Cajaninstilbene acid relaxes rat renal arteries: roles of Ca2+ antagonism and protein kinase C-dependent mechanism.
Cajaninstilbene acid relaxes rat renal arteries: roles of Ca2+ antagonism and protein kinase C-dependent mechanism.
PloS one (2012-10-12)
Dong-Mei Zhang, Yong Li, Wai San Cheang, Chi Wai Lau, Shun-Ming Lin, Qian-Lan Zhang, Nan Yao, Ying Wang, Xin Wu, Yu Huang, Wen-Cai Ye
ABSTRACT
Cajaninstilbene acid (CSA) is a major active component present in the leaves of Cajanus cajan (L.) Millsp. The present study explores the underlying cellular mechanisms for CSA-induced relaxation in rat renal arteries. Vascular reactivity was examined in arterial rings that were suspended in a Multi Myograph System and the expression of signaling proteins was assessed by Western blotting method. CSA (0.1-10 µM) produced relaxations in rings pre-contracted by phenylephrine, serotonin, 9, 11-dideoxy-9α, 11α-epoxymethanoprostaglandin F(2α) (U46619), and 60 mM KCl. CSA-induced relaxations did not show difference between genders and were unaffected by endothelium denudation, nor by treatment with N(G)-nitro-L-arginine methyl ester, indomethacin, ICI-182780, tetraethylammonium ion, BaCl(2), glibenclamide, 4-aminopyridine or propranolol. CSA reduced contraction induced by CaCl(2) (0.01-5 mM) in Ca(2+)-free 60 mM KCl solution and by 30 nM (-)-Bay K8644 in 15 mM KCl solution. CSA inhibited 60 mM KCl-induced Ca(2+) influx in smooth muscle of renal arteries. In addition, CSA inhibited contraction evoked by phorbol 12-myristate 13-acetate (PMA, protein kinase C agonist) in Ca(2+)-free Krebs solution. Moreover, CSA reduced the U46619- and PMA-induced phosphorylation of myosin light chain (MLC) at Ser19 and myosin phosphatase target subunit 1 (MYPT1) at Thr853 which was associated with vasoconstriction. CSA also lowered the phosphorylation of protein kinase C (PKCδ) at Thr505. In summary, the present results suggest that CSA relaxes renal arteries in vitro via multiple cellular mechanisms involving partial inhibition of calcium entry via nifedipine-sensitive calcium channels, protein kinase C and Rho kinase.
Sigma-Aldrich
Tetraethylammonium hydroxide solution, ~25% in methanol (~1.5 M)
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Supelco
Tetraethylammonium chloride, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetraethylammonium chloride, ≥98% (titration)
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 20 wt. % in H2O
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 35 wt. % in H2O
Sigma-Aldrich
Tetraethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Tetraethylammonium hydroxide solution, ~1.0 M (CH3CH2)4NOH in H2O, electrochemical grade
Sigma-Aldrich
(S)-(−)-Bay K8644, ≥98% (HPLC), solid
Sigma-Aldrich
Tetraethylammonium chloride hydrate
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Tetraethylammonium iodide, 98%
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder