Merck
Amphoteric amylopectin: a novel polymeric flocculant.
Carbohydrate polymers (2012-10-10)
Ram Prakash Singh, Sagar Pal, Vijay Kumar Rana, Soumitra Ghorai
ABSTRACT
Novel flocculant based on amphoteric amylopectin for wastewater and industrial effluents treatment has been developed in authors' laboratory. Amphoteric flocculants have anionic and cationic moieties on the same macromolecule and is used to remove both positively and negatively charged contaminant particles in suspensions. Amylopectin based flocculants have been found to be highly efficient flocculant and hence it has been chosen as base polysaccharide. By grafting of polyacrylamide and subsequent hydrolysis, anionic amylopectin has been synthesized. Afterwards, a cationic moiety has been inserted both by chemical/microwave processing. The flocculation efficiency of the amphoteric amylopectin has been tested in kaolin and iron ore suspensions. The results indicate its high efficiency in comparison with anionic, grafted, and base amylopectin. The amphoteric amylopectin prepared via microwave irradiation showed best flocculation efficiency.
Sigma-Aldrich
Amylopectin from potato starch
Sigma-Aldrich
Amylopectin from maize
Sigma-Aldrich
Kaolin, tested according to Ph. Eur.
Sigma-Aldrich
Halloysite nanoclay
Sigma-Aldrich
Kaolin, heavy, powder
Sigma-Aldrich
Kaolin, meets USP testing specifications
Sigma-Aldrich
Kaolin